missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzenesulfonyl chloride, 97%

Catalog No. aaa1264106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1264106 5 g
AAA1264103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1264106 Supplier Thermo Scientific Chemicals Supplier No. A1264106
Only null left

CAS: 445-05-6 | C7H6ClFO2S | 208.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 445-05-6
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.63
MDL Number MFCD00052958
InChI Key NPEWBMJAUUSBLE-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylbenzene-1-sulfonyl chloride, 5-fluoro-2-methylbenzenesulphonyl chloride, 5-fluoro-2-methylbenzenesulfonylchloride, 5-fluoro-2-methyl-benzenesulfonyl chloride, chloro 5-fluoro-2-methylphenyl sulfone, benzenesulfonyl chloride, 5-fluoro-2-methyl, 2-methyl-5-fluorobenzenesulfonyl chloride, pubchem5165, acmc-20ai7v, 5-fluoro-2-methylphenylsulfonyl chloride
PubChem CID 136286
IUPAC Name 5-fluoro-2-methylbenzenesulfonyl chloride
SMILES CC1=CC=C(F)C=C1S(Cl)(=O)=O

Specifications

Density 1.43
Boiling Point 130°C to 132°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 5 g
UN Number UN3265
Beilstein 2643541
Sensitivity Moisture sensitive
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methylbenzenesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only