accessibility menu, dialog, popup

UserName

CAS RN 445-05-6

IUPAC Name: 5-fluoro-2-methylbenzene-1-sulfonyl chloride
Synonyms: 5-fluoro-2-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 208.63
Molecular Formula: C7H6ClFO2S
InChi Key: NPEWBMJAUUSBLE-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1S(Cl)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)