missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylanisole, 98%

Catalog No. p-7028359
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2562903 1 g
AAB2562906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2562903 Supplier Thermo Scientific Chemicals Supplier No. B2562903

CAS: 95729-22-9 | C8H9FO | 140.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95729-22-9
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.157
MDL Number MFCD00673008
InChI Key CXTIUQIZWCABPV-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylanisole, 4-fluoro-2-methoxytoluene, 4-fluoro-2-methoxy-1-methyl-benzene, pubchem2624, 1-methyl-2-methoxy-4-fluorobenzene, 5-fluoro-2-methylphenyl methyl ether, 4-fluoranyl-2-methoxy-1-methyl-benzene, benzene, 4-fluoro-2-methoxy-1-methyl
PubChem CID 2774573
IUPAC Name 4-fluoro-2-methoxy-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)OC

Specifications

Refractive Index 1.494
Quantity 1 g
Formula Weight 140.16
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methylanisole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only