missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylaniline 98.0+%, TCI America™

Catalog No. p-7129558
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
F02325G 5 g
F023225G 25 g
2 options

Catalog No. F02325G

Supplier: TCI America F02325G

Only null left

Chemical Identifiers

CAS 367-29-3
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007764
InChI Key JLCDTNNLXUMYFQ-UHFFFAOYSA-N
Synonym 2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
PubChem CID 67774
IUPAC Name 5-fluoro-2-methylaniline
SMILES CC1=CC=C(F)C=C1N

Specifications

Boiling Point 83°C
Quantity 5 g
Formula Weight 125.15
Percent Purity ≥98.0% (GC)
Chemical Name or Material 5-Fluoro-2-methylaniline