missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxybenzyl bromide, 98+%

Catalog No. AAH2595703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2595703 1 g
AAH2595706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2595703 Supplier Thermo Scientific Chemicals Supplier No. H2595703
Only null left

CAS: 700381-18-6 | C8H8BrFO | 219.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 700381-18-6
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.05
MDL Number MFCD00671769
InChI Key LXUGHXUXEMUEKR-UHFFFAOYSA-N
Synonym 2-bromomethyl-4-fluoro-1-methoxybenzene, 5-fluoro-2-methoxybenzyl bromide, 2-methyloxy-5-fluorobenzyl bromide, 5-fluoro-2-methyloxybenzyl bromide, alpha-bromo-5-fluoro-2-methoxytoluene, pubchem4924, 2-bromomethyl-4-fluoroanisole, ablock ab-14-0063, 2-bromomethyl-4-fluoro-1-methoxy-benzene, 5-fluoro-2-methoxybenzyl bromide 97
PubChem CID 20111745
IUPAC Name 2-(bromomethyl)-4-fluoro-1-methoxybenzene
SMILES COC1=C(CBr)C=C(F)C=C1

Specifications

Melting Point 58°C to 62°C
Quantity 1 g
UN Number UN3261
Formula Weight 219.05
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2-methoxybenzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only