missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxybenzaldehyde, 98%

Catalog No. AAB2468403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2468403 1 g
AAB2468406 5 g
AAB2468414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2468403 Supplier Thermo Scientific Chemicals Supplier No. B2468403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19415-51-1 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19415-51-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143458
InChI Key CRLDWFVRQNUUSZ-UHFFFAOYSA-N
PubChem CID 2734943
IUPAC Name 5-fluoro-2-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)F)C=O

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Synonym 2-methoxy-5-fluorobenzaldehyde, 5-fluoro-o-anisaldehyde, 5-fluoro-o-methoxybenzaldehyde, benzaldehyde, 5-fluoro-2-methoxy, 5-fluoro-2-methoxy-benzaldehyde, 5-fluoro-o-2-methoxybenzaldehyde, pubchem2636, acmc-209exz, 2-methoxy-5-fluoro benzaldehyde, benzaldehyde,5-fluoro-2-methoxy
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only