missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxy-4-pyrimidinone 98.0+%, TCI America™

Catalog No. p-7129746
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05475G 5 g
F054725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05475G Supplier TCI America Supplier No. F05475G

Chemical Identifiers

CAS 1480-96-2
Molecular Formula C5H5FN2O2
Molecular Weight (g/mol) 144.105
InChI Key VMIFBCPINLZNNI-UHFFFAOYSA-N
PubChem CID 352550
IUPAC Name 5-fluoro-2-methoxy-1H-pyrimidin-6-one
SMILES COC1=NC=C(C(=O)N1)F

Specifications

Melting Point 200°C
Color White
Quantity 5 g
Formula Weight 144.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-methoxy-4-pyrimidinone