accessibility menu, dialog, popup

UserName

H3CONFOHN

CAS RN 1480-96-2

IUPAC Name: 5-fluoro-2-methoxy-3,4-dihydropyrimidin-4-one
Synonyms: 5-fluoro-2-methoxy-3H-pyrimidin-4-one
Molecular Weight (g/mol): 144.11
Molecular Formula: C5H5FN2O2
InChi Key: VMIFBCPINLZNNI-UHFFFAOYSA-N
SMILES: COC1=NC=C(F)C(=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results