missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-iodoaniline, 97%

Catalog No. AAL1772306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1772306 5 g
AAL1772303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1772306 Supplier Thermo Scientific Chemicals Supplier No. L1772306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 255724-71-1 | C6H5FIN | 237.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 255724-71-1
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.02
MDL Number MFCD02093953
InChI Key UCPDOOZBROQHME-UHFFFAOYSA-N
Synonym benzenamine, 5-fluoro-2-iodo, 5-fluoro-2-iodo-aniline, 5-fluoro-2-iodo-phenylamine, 2-iodo-5-fluoroaniline, 5-fluoro-2-iodophenylamine, pubchem1535, acmc-20a2iw, 3-fluoro-6-iodoaniline, intermediates-zcf02682, 5-fluoro-2-iodobenzenamine
PubChem CID 2774512
IUPAC Name 5-fluoro-2-iodoaniline
SMILES NC1=C(I)C=CC(F)=C1

Specifications

Melting Point 40°C to 42°C
Quantity 5 g
UN Number UN2811
Beilstein 8478850
Sensitivity Light sensitive
Formula Weight 237.02
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-iodoaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only