missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (+)-5-Fluoro-2'-deoxyuridine, 99+%

Catalog No. AC227601000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227601000 100 mg Glass bottle
AC227605000 500 mg Glass bottle
AC227600025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227601000 Supplier Thermo Scientific Chemicals Supplier No. 227601000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-91-9
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00006530
InChI Key ODKNJVUHOIMIIZ-RRKCRQDMSA-N
Synonym floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
PubChem CID 5790
ChEBI CHEBI:60761
IUPAC Name 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Melting Point 148.0°C to 153.0°C
Color White
Loss on Drying 0.5% (60°C, 4 hrs) (vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15,4144
Specific Rotation Condition + 35.90 (20.00°C c=1,water)
Specific Rotation + 35.90
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,practically insoluble in chcl3,insoluble in ether and hexane
Formula Weight 246.19
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material (+)-5-Fluoro-2′-deoxyuridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
IF ON SKIN: Wash with plenty of soap and
EINECSNumber 200-072-5

RUO – Research Use Only