missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2'-deoxyuridine, 98+%

Catalog No. AAL16497ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL16497ME 500 mg
AAL16497MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL16497ME Supplier Thermo Scientific Chemicals Supplier No. L16497ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol

5-Fluoro-2'-deoxyuridine acts as an antiviral and antineoplastic agent. It is also employed for renal function diagnosis. Further, it is used as an experimental anticancer agent, which shows activity against a variety of malignant neoplasms. It is also used as an inhibitor of thymidylate synthetase and DNA synthesis. In addition to this, it plays an important role in the treatment of colorectal cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol, isopropyl alcohol, methanol, aqueous buffers, acetone, dimethyl sulfoxide, and ethanol. Insoluble in ether, benzene and chloroform.

Notes
Store in a cool place.Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 50-91-9
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00006530
InChI Key ODKNJVUHOIMIIZ-RRKCRQDMSA-N
Synonym floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
PubChem CID 5790
ChEBI CHEBI:60761
IUPAC Name 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Melting Point 148°C to 151°C
Quantity 500 mg
UN Number UN2811
Beilstein 2645818
Merck Index 14,4112
Solubility Information Soluble in water,alcohol,isopropyl alcohol,methanol,aqueous buffers,acetone,dimethyl sulfoxide,and ethanol. Insoluble in ether,benzene and chloroform.
Formula Weight 246.2
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2'-deoxyuridine
Hazard Category H301-H315-H319-H341
Hazard Statement GHS H Statement
H360-H341-H302
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-072-5
RTECSNumber YU7535000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only