missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-1-(tetrahydro-2-furfuryl)uracil 98.0+%, TCI America™

Catalog No. p-7129798
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06355G 5 g
F063525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06355G Supplier TCI America Supplier No. F06355G

Chemical Identifiers

CAS 17902-23-7
Molecular Formula C8H9FN2O3
Molecular Weight (g/mol) 200.17
MDL Number MFCD00012351
InChI Key WFWLQNSHRPWKFK-UHFFFAOYNA-N
Synonym tegafur, ftorafur, futraful, fluorofur, sinoflurol, citofur, fental, neberk, coparogin, florafur
PubChem CID 5386
ChEBI CHEBI:32188
IUPAC Name 5-fluoro-1-(oxolan-2-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES FC1=CN(C2CCCO2)C(=O)NC1=O

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 200.17
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-1-(tetrahydro-2-furfuryl)uracil
EINECSNumber (9)-0855
RTECSNumber YR0450000
TSCA Yes