CAS RN 17902-23-7
IUPAC Name:
5-fluoro-1-(oxolan-2-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
tegafur
Molecular Weight (g/mol):
200.17
Molecular Formula:
C8H9FN2O3
InChi Key:
WFWLQNSHRPWKFK-UHFFFAOYNA-N
SMILES:
FC1=CN(C2CCCO2)C(=O)NC1=O
1
–
1
of
1
results
1
5-Fluoro-1-(tetrahydro-2-furfuryl)uracil 98.0+%, TCI America™
CAS: 17902-23-7 Molecular Formula: C8H9FN2O3 Molecular Weight (g/mol): 200.17 MDL Number: MFCD00012351 InChI Key: WFWLQNSHRPWKFK-UHFFFAOYNA-N Synonym: tegafur,ftorafur,futraful,fluorofur,sinoflurol,citofur,fental,neberk,coparogin,florafur PubChem CID: 5386 ChEBI: CHEBI:32188 IUPAC Name: 5-fluoro-1-(oxolan-2-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione SMILES: FC1=CN(C2CCCO2)C(=O)NC1=O