missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-1-indanone 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 37°C |
| Color | White-Yellow |
| Boiling Point | 114°C |
| Quantity | 5 g |
| Formula Weight | 150.15 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Fluoro-1-indanone |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |