accessibility menu, dialog, popup

UserName

FO

CAS RN 700-84-5

IUPAC Name: 5-fluoro-2,3-dihydro-1H-inden-1-one
Synonyms: 5-fluoro-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 150.15
Molecular Formula: C9H7FO
InChi Key: WVPPBVAMKNQXJA-UHFFFAOYSA-N
SMILES: FC1=CC=C2C(=O)CCC2=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)