missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-1,3-dimethyl-2-nitrobenzene, 98%

Catalog No. AAH2716603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2716603 1 g
AAH2716606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2716603 Supplier Thermo Scientific Chemicals Supplier No. H2716603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 315-12-8 | C8H8FNO2 | 169.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 315-12-8
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD03413407
InChI Key KDPQDERPJHLWGF-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-fluoronitrobenzene, 5-fluoro-2-nitro-m-xylene, 1,3-dimethyl-5-fluoro-2-nitrobenzene, benzene, 5-fluoro-1,3-dimethyl-2-nitro, 4-fluoro-2,6-dimethyl-1-nitrobenzene, benzene,5-fluoro-1,3-dimethyl-2-nitro, 5-fluoro-2-nitro-1,3-dimethylbenzene
PubChem CID 12651353
IUPAC Name 5-fluoro-1,3-dimethyl-2-nitrobenzene
SMILES CC1=CC(=CC(=C1[N+](=O)[O-])C)F

Specifications

Melting Point 52°C to 57°C
Quantity 1 g
Formula Weight 169.16
Percent Purity 98%
Chemical Name or Material 5-Fluoro-1,3-dimethyl-2-nitrobenzene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only