missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Ethynyl-2'-deoxycytidine 98.0+%, TCI America™
$1164.72 - $1164.72
Chemical Identifiers
| CAS | 69075-47-4 |
|---|---|
| Molecular Formula | C11H13N3O4 |
| Molecular Weight (g/mol) | 251.242 |
| MDL Number | MFCD22417249 |
| InChI Key | HMJSYXIPNSASJY-IWSPIJDZSA-N |
| Synonym | EdC |
| PubChem CID | 90176383 |
| IUPAC Name | 4-amino-5-ethynyl-1-[(2R,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| SMILES | C#CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E1093200MG
|
TCI America
E1093200MG |
200 mg |
Each for $1,164.72
|
|
|||||
Chemical Identifiers
| 69075-47-4 | |
| 251.242 | |
| HMJSYXIPNSASJY-IWSPIJDZSA-N | |
| 90176383 | |
| C#CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O |
| C11H13N3O4 | |
| MFCD22417249 | |
| EdC | |
| 4-amino-5-ethynyl-1-[(2R,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
Specifications
| 69075-47-4 | |
| C11H13N3O4 | |
| 200 mg | |
| HMJSYXIPNSASJY-IWSPIJDZSA-N | |
| 4-amino-5-ethynyl-1-[(2R,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one | |
| 90176383 | |
| ≥98.0% (N) | |
| 5-Ethynyl-2′-deoxycytidine |
| 190°C | |
| MFCD22417249 | |
| EdC | |
| C#CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O | |
| 251.242 | |
| 251.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : No