missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyluracil 98.0+%, TCI America™

Catalog No. p-7129164
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08071G 1 g
E08075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08071G Supplier TCI America Supplier No. E08071G

Chemical Identifiers

CAS 4212-49-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00079187
InChI Key RHIULBJJKFDJPR-UHFFFAOYSA-N
Synonym 5-ethyluracil, 5-ethyl uracil, 2,4-dihydroxy-5-ethylpyrimidine, 5-ethylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 5-ethyl, 5-ethyl-1,3-dihydropyrimidine-2,4-dione, 5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, uracil, 5-ethyl, zlchem 329, acmc-209jno
PubChem CID 73267
IUPAC Name 5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CCC1=CNC(=O)NC1=O

Specifications

Melting Point 303°C
Color White-Yellow
Quantity 1 g
Formula Weight 140.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Ethyluracil