CAS RN 4212-49-1
CAS RN 4212-49-1
IUPAC Name:
5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
5-ethyl-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol):
140.14
Molecular Formula:
C6H8N2O2
InChi Key:
RHIULBJJKFDJPR-UHFFFAOYSA-N
SMILES:
CCC1=CNC(=O)NC1=O
1
–
1
of
1
results
1
5-Ethyluracil 98.0+%, TCI America™
CAS: 4212-49-1 Molecular Formula: C6H8N2O2 Molecular Weight (g/mol): 140.14 MDL Number: MFCD00079187 InChI Key: RHIULBJJKFDJPR-UHFFFAOYSA-N Synonym: 5-ethyluracil,5-ethyl uracil,2,4-dihydroxy-5-ethylpyrimidine,5-ethylpyrimidine-2,4-diol,2,4 1h,3h-pyrimidinedione, 5-ethyl,5-ethyl-1,3-dihydropyrimidine-2,4-dione,5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione,uracil, 5-ethyl,zlchem 329,acmc-209jno PubChem CID: 73267 IUPAC Name: 5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione SMILES: CCC1=CNC(=O)NC1=O