missing translation for 'onlineSavingsMsg'
Learn More

5-Ethylpyridine-2,3-dicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129202
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08875G 5 g
E088725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08875G Supplier TCI America Supplier No. E08875G

Chemical Identifiers

CAS 102268-15-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00071733
InChI Key MTAVBTGOXNGCJR-UHFFFAOYSA-N
Synonym 5-ethylquinolinic acid, 5-ethyl-2,3-pyridinedicarboxylic acid, 2,3-pyridinedicarboxylic acid, 5-ethyl, 5-ethyl-2,3-pyridine dicarboxylic acid, 5-ethylpyridine-2,3-dicarboxylicacid, pubchem16905, dsstox_cid_9072, dsstox_rid_78667, dsstox_gsid_29072, ksc179a3d
PubChem CID 113606
IUPAC Name 5-ethylpyridine-2,3-dicarboxylic acid
SMILES CCC1=CN=C(C(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 156°C
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Ethylpyridine-2,3-dicarboxylic Acid
EINECSNumber (5)-6175
RTECSNumber US8005000
TSCA Yes