missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Ethoxy-2-mercaptobenzimidazole 98.0+%, TCI America™
$221.44 - $827.08
Chemical Identifiers
| CAS | 55489-15-1 |
|---|---|
| Molecular Formula | C9H10N2OS |
| Molecular Weight (g/mol) | 194.252 |
| MDL Number | MFCD01326487 |
| InChI Key | WUSCBOFBIYZVCQ-UHFFFAOYSA-N |
| Synonym | 5-ethoxy-2-mercaptobenzimidazole, 5-ethoxy-1h-benzo d imidazole-2-thiol, 5-ethoxy-1h-benzimidazole-2-thiol, 5-ethoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-ethoxybenzimidazole, 5-ethoxybenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-ethoxy-1,3-dihydro, acmc-1arr4, ksc603o0j, 5-ethoxy-2-benzimidazolethiol |
| PubChem CID | 790793 |
| IUPAC Name | 5-ethoxy-1,3-dihydrobenzimidazole-2-thione |
| SMILES | CCOC1=CC2=C(C=C1)NC(=S)N2 |
Chemical Identifiers
| 55489-15-1 | |
| 194.252 | |
| WUSCBOFBIYZVCQ-UHFFFAOYSA-N | |
| 790793 | |
| CCOC1=CC2=C(C=C1)NC(=S)N2 |
| C9H10N2OS | |
| MFCD01326487 | |
| 5-ethoxy-2-mercaptobenzimidazole, 5-ethoxy-1h-benzo d imidazole-2-thiol, 5-ethoxy-1h-benzimidazole-2-thiol, 5-ethoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-ethoxybenzimidazole, 5-ethoxybenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-ethoxy-1,3-dihydro, acmc-1arr4, ksc603o0j, 5-ethoxy-2-benzimidazolethiol | |
| 5-ethoxy-1,3-dihydrobenzimidazole-2-thione |
Specifications
| 55489-15-1 | |
| C9H10N2OS | |
| 5 g | |
| WUSCBOFBIYZVCQ-UHFFFAOYSA-N | |
| 5-ethoxy-1,3-dihydrobenzimidazole-2-thione | |
| 790793 | |
| ≥98.0% (HPLC) | |
| 5-Ethoxy-2-mercaptobenzimidazole |
| 246°C | |
| MFCD01326487 | |
| 5-ethoxy-2-mercaptobenzimidazole, 5-ethoxy-1h-benzo d imidazole-2-thiol, 5-ethoxy-1h-benzimidazole-2-thiol, 5-ethoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-ethoxybenzimidazole, 5-ethoxybenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-ethoxy-1,3-dihydro, acmc-1arr4, ksc603o0j, 5-ethoxy-2-benzimidazolethiol | |
| CCOC1=CC2=C(C=C1)NC(=S)N2 | |
| 194.252 | |
| 194.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : No