missing translation for 'onlineSavingsMsg'
Learn More

5-Ethoxy-2-mercaptobenzimidazole 98.0+%, TCI America™

Supplier:  TCI America E053525G

 View more versions of this product

Catalog No. E053525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Ethoxy-2-mercaptobenzimidazole
246°C
MFCD01326487
5-ethoxy-2-mercaptobenzimidazole, 5-ethoxy-1h-benzo d imidazole-2-thiol, 5-ethoxy-1h-benzimidazole-2-thiol, 5-ethoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-ethoxybenzimidazole, 5-ethoxybenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-ethoxy-1,3-dihydro, acmc-1arr4, ksc603o0j, 5-ethoxy-2-benzimidazolethiol
CCOC1=CC2=C(C=C1)NC(=S)N2
194.252
194.25
Crystalline Powder
55489-15-1
C9H10N2OS
25 g
WUSCBOFBIYZVCQ-UHFFFAOYSA-N
5-ethoxy-1,3-dihydrobenzimidazole-2-thione
790793
≥98.0% (HPLC)

Chemical Identifiers

55489-15-1
194.252
WUSCBOFBIYZVCQ-UHFFFAOYSA-N
790793
CCOC1=CC2=C(C=C1)NC(=S)N2
C9H10N2OS
MFCD01326487
5-ethoxy-2-mercaptobenzimidazole, 5-ethoxy-1h-benzo d imidazole-2-thiol, 5-ethoxy-1h-benzimidazole-2-thiol, 5-ethoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-ethoxybenzimidazole, 5-ethoxybenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-ethoxy-1,3-dihydro, acmc-1arr4, ksc603o0j, 5-ethoxy-2-benzimidazolethiol
5-ethoxy-1,3-dihydrobenzimidazole-2-thione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions