missing translation for 'onlineSavingsMsg'
Learn More

5-Dimethylamino-2-nitrosophenol Hydrochloride 98.0+%, TCI America™

Catalog No. p-7133711
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N02615G Supplier TCI America Supplier No. N02615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41317-10-6
Molecular Formula C8H11ClN2O2
Molecular Weight (g/mol) 202.638
MDL Number MFCD00060162
InChI Key WRIZNEWVHJJMHU-UHFFFAOYSA-N
Synonym 2-Nitroso-5-dimethylaminophenol Hydrochloride
PubChem CID 9815614
IUPAC Name 5-(dimethylamino)-2-nitrosophenol;hydrochloride
SMILES CN(C)C1=CC(=C(C=C1)N=O)O.Cl

Specifications

Quantity 5 g
Formula Weight 202.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Dimethylamino-2-nitrosophenol Hydrochloride