accessibility menu, dialog, popup

UserName

CAS RN 41317-10-6

IUPAC Name: 5-(dimethylamino)-2-nitrosophenol hydrochloride
Synonyms: 5-(dimethylamino)-2-nitrosophenol hydrochloride
Molecular Weight (g/mol): 202.64
Molecular Formula: C8H11ClN2O2
InChi Key: WRIZNEWVHJJMHU-UHFFFAOYSA-N
SMILES: Cl.CN(C)C1=CC(O)=C(C=C1)N=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results