missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanothiophene-2-carboxaldehyde, 95%

Catalog No. p-7055794
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6446203 1 g
AAH6446206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6446203 Supplier Thermo Scientific Chemicals Supplier No. H6446203
Only null left

CAS: 21512-16-3 | C6H3NOS | 137.156 g/mol

5-Cyanothiophene-2-carboxaldehyde is found in caper and may have pest control activity against meloidogyne incognita.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Cyanothiophene-2-carboxaldehyde is found in caper and may have pest control activity against meloidogyne incognita.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 21512-16-3
Molecular Formula C6H3NOS
Molecular Weight (g/mol) 137.156
MDL Number MFCD09037804
InChI Key PZIFYWVUYHMYOA-UHFFFAOYSA-N
Synonym 5-cyano-2-thiophene carbaldehyde, 5-cyano-2-thiophenecarboxaldehyde, 5-cyanothiophene-2-carboxaldehyde, 2-thiophenecarbonitrile, 5-formyl, 5-cyano-2-thiophenecarbaldehyde, 2-formyl-5-cyanothiophen, 2-cyano-5-formylthiophene, 2-formyl-5-cyanothiophene, 2-cyano-5-formyl-thiophene, 5-cyanothiophene-2-aldehyde
PubChem CID 12280004
IUPAC Name 5-formylthiophene-2-carbonitrile
SMILES C1=C(SC(=C1)C#N)C=O

Specifications

Melting Point 90°C to 95°C
Color White
Quantity 1 g
UN Number UN3439
Beilstein 115517
Solubility Information Slightly soluble in water.
Formula Weight 137.16
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Cyanothiophene-2-carboxaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only