missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanopyridine-2-carboxylic acid, 95%

CAS: 53234-55-2 | C7H4N2O2 | 148.12 g/mol

$112.81 - $229.49

Chemical Identifiers

CAS 53234-55-2
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD09152775
InChI Key HRLVPHGCEGTVLK-UHFFFAOYSA-N
Synonym 5-cyanopicolinic acid, 5-cyano-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-cyano, 5-cyano-2-pyridincarboxylic acid, 5-cyanopicolinicacid, 5-cyano-picolinic acid, pubchem16740, acmc-1cubt, 5-cyano-2-picolinic acid, 5-cyano-2-pyridine carboxylic acid
PubChem CID 12400734
IUPAC Name 5-cyanopyridine-2-carboxylic acid
SMILES OC(=O)C1=NC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH33722MD
View Documents
Thermo Scientific Chemicals
H33722MD
250 mg
Each for $112.81
Only null left
 
AAH3372203
View Documents
Thermo Scientific Chemicals
H3372203
1 g
Each for $229.49
Only null left
 
Description

Description

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

53234-55-2
148.12
HRLVPHGCEGTVLK-UHFFFAOYSA-N
12400734
OC(=O)C1=NC=C(C=C1)C#N
C7H4N2O2
MFCD09152775
5-cyanopicolinic acid, 5-cyano-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-cyano, 5-cyano-2-pyridincarboxylic acid, 5-cyanopicolinicacid, 5-cyano-picolinic acid, pubchem16740, acmc-1cubt, 5-cyano-2-picolinic acid, 5-cyano-2-pyridine carboxylic acid
5-cyanopyridine-2-carboxylic acid

Specifications

53234-55-2
C7H4N2O2
250 mg
5-cyanopicolinic acid, 5-cyano-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-cyano, 5-cyano-2-pyridincarboxylic acid, 5-cyanopicolinicacid, 5-cyano-picolinic acid, pubchem16740, acmc-1cubt, 5-cyano-2-picolinic acid, 5-cyano-2-pyridine carboxylic acid
HRLVPHGCEGTVLK-UHFFFAOYSA-N
5-cyanopyridine-2-carboxylic acid
12400734
95%
212°C to 214°C
MFCD09152775
UN3439
Sparingly soluble in water.(0.26g/L) (25°C),
OC(=O)C1=NC=C(C=C1)C#N
148.12
148.1
5-Cyanopyridine-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only