missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanopyridine-2-carboxylic acid, 95%

CAS: 53234-55-2 | C7H4N2O2 | 148.12 g/mol

Supplier:  Thermo Scientific Chemicals H3372203

 View more versions of this product

Catalog No. AAH3372203


Only null left
Explore available promotions

Description

Description

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Specifications

5-Cyanopyridine-2-carboxylic acid
212°C to 214°C
MFCD09152775
UN3439
Sparingly soluble in water.(0.26g/L) (25°C),
OC(=O)C1=NC=C(C=C1)C#N
148.12
148.1
53234-55-2
C7H4N2O2
1 g
5-cyanopicolinic acid, 5-cyano-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-cyano, 5-cyano-2-pyridincarboxylic acid, 5-cyanopicolinicacid, 5-cyano-picolinic acid, pubchem16740, acmc-1cubt, 5-cyano-2-picolinic acid, 5-cyano-2-pyridine carboxylic acid
HRLVPHGCEGTVLK-UHFFFAOYSA-N
5-cyanopyridine-2-carboxylic acid
12400734
95%

Chemical Identifiers

53234-55-2
148.12
HRLVPHGCEGTVLK-UHFFFAOYSA-N
12400734
OC(=O)C1=NC=C(C=C1)C#N
C7H4N2O2
MFCD09152775
5-cyanopicolinic acid, 5-cyano-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-cyano, 5-cyano-2-pyridincarboxylic acid, 5-cyanopicolinicacid, 5-cyano-picolinic acid, pubchem16740, acmc-1cubt, 5-cyano-2-picolinic acid, 5-cyano-2-pyridine carboxylic acid
5-cyanopyridine-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only