missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanophthalide 98.0+%, TCI America™

Catalog No. C20015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20015G 5 g
C200125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20015G Supplier TCI America Supplier No. C20015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82104-74-3
Molecular Formula C9H5NO2
Molecular Weight (g/mol) 159.144
InChI Key XEEGWTLAFIZLSF-UHFFFAOYSA-N
Synonym 5-cyanophthalide, 1-oxo-1,3-dihydroisobenzofuran-5-carbonitrile, 5-cyano phthalide, 5-cyano-3h-isobenzofuranone, 1,3-dihydro-1-oxoisobenzofuran-5-carbonitrile, 1-oxo-1,3-dihydro-2-benzofuran-5-carbonitrile, 1-oxo-3h-isobenzofuran-5-carbonitrile, 1,3-dihydro-1-oxo-5-isobenzofurancarbonitrile, 5-isobenzofurancarbonitrile, 1,3-dihydro-1-oxo, 5-cyanophthalid
PubChem CID 821218
IUPAC Name 1-oxo-3H-2-benzofuran-5-carbonitrile
SMILES C1C2=C(C=CC(=C2)C#N)C(=O)O1

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 5 g
Formula Weight 159.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyanophthalide