accessibility menu, dialog, popup

UserName

CAS RN 82104-74-3

IUPAC Name: 1-oxo-1,3-dihydro-2-benzofuran-5-carbonitrile
Synonyms: 1-oxo-3H-2-benzofuran-5-carbonitrile
Molecular Weight (g/mol): 159.14
Molecular Formula: C9H5NO2
InChi Key: XEEGWTLAFIZLSF-UHFFFAOYSA-N
SMILES: O=C1OCC2=CC(=CC=C12)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results