missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanoindole 98.0+%, TCI America™

Catalog No. C14911G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14911G 1 g
C14915G 5 g
C149125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C14911G Supplier TCI America Supplier No. C14911G

Chemical Identifiers

CAS 15861-24-2
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00005669
InChI Key YHYLDEVWYOFIJK-UHFFFAOYSA-N
Synonym 5-cyanoindole, indole-5-carbonitrile, 5-cyano indole, 5-cyano-1h-indole, indole-5-cyano, 5-indolecarbonitrile, 5-cyano-indole, indol-5-carbonitrile, indole, 5-cyano, pubchem7326
PubChem CID 27513
IUPAC Name 1H-indole-5-carbonitrile
SMILES C1=CC2=C(C=CN2)C=C1C#N

Specifications

Melting Point 106°C
Quantity 1 g
Formula Weight 142.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyanoindole
RTECSNumber NL5993120
TSCA No