missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-fluoropyridine, 95%

Catalog No. AAH6427803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6427803 1 g
AAH6427806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6427803 Supplier Thermo Scientific Chemicals Supplier No. H6427803
Only null left

CAS: 3939-12-6 | C7H8BrN | 186.05 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 3939-12-6
MDL Number MFCD09261087

Specifications

Melting Point 51°C to 52°C
Color White
Molecular Formula C7H8BrN
Quantity 1 g
UN Number UN3439
Beilstein 471855
Synonym 2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
Solubility Information Slightly soluble in water.
InChI Key NOYASZMZIBFFNZ-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)CN)Br
IUPAC Name (2-bromophenyl)methanamine
Molecular Weight (g/mol) 186.05
PubChem CID 334072
Formula Weight 122.1
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Cyano-2-fluoropyridine
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only