accessibility menu, dialog, popup

UserName

CAS RN 3939-12-6

NNF

CAS RN 3939-12-6

IUPAC Name: 6-fluoropyridine-3-carbonitrile
Synonyms: 6-fluoropyridine-3-carbonitrile
Molecular Weight (g/mol): 122.1
Molecular Formula: C6H3FN2
InChi Key: VRLVOMUVHHHJHB-UHFFFAOYSA-N
SMILES: FC1=NC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results