missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-1H-imidazole-4-carboxamide Hydrate 95.0+%, TCI America™

Catalog No. C13385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13385G 5 g
C133825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13385G Supplier TCI America Supplier No. C13385G

Chemical Identifiers

CAS 5372-23-6
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00129664
InChI Key QBKYWNHLILCGEI-UHFFFAOYSA-N
Synonym 5-cyano-1h-imidazole-4-carboxamide, 4-cyano-5-imidazolecarboxamide, 5-cyano-3h-imidazole-4-carboxamide, 1h-imidazole-4-carboxamide, 5-cyano, 4-cyanoimidazole-5-carboxamide, acmc-20amkb, 5-cyanoimidazole-4-carboxamide, 1h-imidazole-5-carboxamide,4-cyano, 5-cyano-1h-imidazole-4-carboxamidehydrate, 1h-imidazole-4-carboxamide, 5-cyano-9ci
PubChem CID 270862
IUPAC Name 4-cyano-1H-imidazole-5-carboxamide
SMILES NC(=O)C1=C(N=CN1)C#N

Specifications

Melting Point 278°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 136.11
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyano-1H-imidazole-4-carboxamide Hydrate