accessibility menu, dialog, popup

UserName

NH2ONHNN

CAS RN 5372-23-6

IUPAC Name: 4-cyano-1H-imidazole-5-carboxamide
Synonyms: 5-cyano-3H-imidazole-4-carboxamide
Molecular Weight (g/mol): 136.11
Molecular Formula: C5H4N4O
InChi Key: QBKYWNHLILCGEI-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N=CN1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results