missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-CR 6G, SE (5-Carboxyrhodamine 6G, Succinimidyl Ester), single isomer

Catalog No. C6127
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C6127 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C6127 Supplier Invitrogen™ Supplier No. C6127

5-CR 6G, SE (5-Carboxyrhodamine 6G, Succinimidyl Ester), single isomer

The amine-reactive form of 5-CR 6G is used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lisaamine™ rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red™ conjugates.

Specifications

Quantity 5 mg
Product Type 5-CR 6G SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Rhodamine 6G
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.