missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovanillic acid, tech. 90%

Catalog No. p-7020774
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1284106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1284106 Supplier Thermo Scientific Chemicals Supplier No. A1284106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62936-23-6 | C8H7ClO4 | 202.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62936-23-6
Molecular Formula C8H7ClO4
Molecular Weight (g/mol) 202.59
MDL Number MFCD00016531
InChI Key XBRYEHVBBMSSCG-UHFFFAOYSA-N
PubChem CID 44215
IUPAC Name 3-chloro-4-hydroxy-5-methoxybenzoic acid
SMILES COC1=C(C(=CC(=C1)C(=O)O)Cl)O

Specifications

Melting Point 242°C to 248°C
Quantity 5 g
Beilstein 2648897
Synonym 5-chlorovanillic acid, 3-chloro-4-hydroxy-5-methoxy-benzoic acid, benzoic acid, 3-chloro-4-hydroxy-5-methoxy, 3-chloro-4-hydroxy-5-methoxy benzoic acid, 5-chlorovanillicacid, 5-chloro-vanillic acid, acmc-1b2hg, chloro-4-hydroxy-3-methoxybenzoic acid, 5-chloro-4-hydroxy-3-methoxybenzoic acid, benzoic acid,3-chloro-4-hydroxy-5-methoxy
Formula Weight 202.6
Percent Purity 90%
Grade Technical
Chemical Name or Material 5-Chlorovanillic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 263-766-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only