missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chlorovaleric Acid 96.0+%, TCI America™
$37.29 - $82.02
Chemical Identifiers
| CAS | 1119-46-6 |
|---|---|
| Molecular Formula | C5H9ClO2 |
| Molecular Weight (g/mol) | 136.58 |
| MDL Number | MFCD00004415 |
| InChI Key | YSXDKDWNIPOSMF-UHFFFAOYSA-N |
| Synonym | 5-Chloropentanoic Acid |
| PubChem CID | 14244 |
| ChEBI | CHEBI:60701 |
| IUPAC Name | 5-chloropentanoic acid |
| SMILES | OC(=O)CCCCCl |
Chemical Identifiers
| 1119-46-6 | |
| 136.58 | |
| YSXDKDWNIPOSMF-UHFFFAOYSA-N | |
| 14244 | |
| 5-chloropentanoic acid |
| C5H9ClO2 | |
| MFCD00004415 | |
| 5-Chloropentanoic Acid | |
| CHEBI:60701 | |
| OC(=O)CCCCCl |
Specifications
| 1119-46-6 | |
| 136°C | |
| MFCD00004415 | |
| 5-Chloropentanoic Acid | |
| OC(=O)CCCCCl | |
| 136.58 | |
| CHEBI:60701 | |
| ≥96.0% (GC,T) | |
| 5-Chlorovaleric Acid |
| Yellow | |
| C5H9ClO2 | |
| 5 g | |
| YSXDKDWNIPOSMF-UHFFFAOYSA-N | |
| 5-chloropentanoic acid | |
| 14244 | |
| 136.58 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1180
TSCA : No