accessibility menu, dialog, popup

UserName

HOOCl

CAS RN 1119-46-6

IUPAC Name: 5-chloropentanoic acid
Synonyms: 5-chlorovaleric acid ω-chlorovaleric acid ω-chloropentanoic acid
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: YSXDKDWNIPOSMF-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results