missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleric Acid 96.0+%, TCI America™

Catalog No. C03095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03095G 5 g
C030925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03095G Supplier TCI America Supplier No. C03095G

Chemical Identifiers

CAS 1119-46-6
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00004415
InChI Key YSXDKDWNIPOSMF-UHFFFAOYSA-N
Synonym 5-Chloropentanoic Acid
PubChem CID 14244
ChEBI CHEBI:60701
IUPAC Name 5-chloropentanoic acid
SMILES OC(=O)CCCCCl

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 136.58
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Chlorovaleric Acid
EINECSNumber (2)-1180
TSCA No