missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-sulfonamide, 97%

CAS: 53595-66-7 | C4H4ClNO2S2 | 197.65 g/mol

Supplier:  Thermo Scientific Chemicals B2047706

 View more versions of this product

Catalog No. AAB2047706


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chlorothiophene-2-sulfonamide
115°C to 118°C
MFCD00052584
5-chloro-2-thiophenesulfonamide, 2-thiophenesulfonamide, 5-chloro, 2-chloro thiophene-5-sulfonamide, pubchem7381, 5-chlorothiophenesulfonamide, 4-18-00-06708 beilstein handbook reference, ksc490i0r, 2-chloro-5-sulfamoylthiophene, 5-chloro-2-thienylsulphonamide
NS(=O)(=O)C1=CC=C(Cl)S1
197.65
197.67
53595-66-7
C4H4ClNO2S2
5 g
RKLQLYBJAZBSEU-UHFFFAOYSA-N
5-chlorothiophene-2-sulfonamide
1241301
97%

Chemical Identifiers

53595-66-7
197.65
RKLQLYBJAZBSEU-UHFFFAOYSA-N
1241301
NS(=O)(=O)C1=CC=C(Cl)S1
C4H4ClNO2S2
MFCD00052584
5-chloro-2-thiophenesulfonamide, 2-thiophenesulfonamide, 5-chloro, 2-chloro thiophene-5-sulfonamide, pubchem7381, 5-chlorothiophenesulfonamide, 4-18-00-06708 beilstein handbook reference, ksc490i0r, 2-chloro-5-sulfamoylthiophene, 5-chloro-2-thienylsulphonamide
5-chlorothiophene-2-sulfonamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only