accessibility menu, dialog, popup

UserName

CAS RN 53595-66-7

IUPAC Name: 5-chlorothiophene-2-sulfonamide
Synonyms: 5-chlorothiophene-2-sulfonamide
Molecular Weight (g/mol): 197.65
Molecular Formula: C4H4ClNO2S2
InChi Key: RKLQLYBJAZBSEU-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(Cl)S1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)