missing translation for 'onlineSavingsMsg'
Learn More

5-(Chlorosulfonyl)salicylic acid, 97%

Catalog No. p-7033282
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5037403 1 g
AAH5037406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5037403 Supplier Thermo Scientific Chemicals Supplier No. H5037403
Only null left

CAS: 17243-13-9 | C7H5ClO5S | 236.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17243-13-9
Molecular Formula C7H5ClO5S
Molecular Weight (g/mol) 236.62
MDL Number MFCD00052974
InChI Key SJXHSFSHNKFRLN-UHFFFAOYSA-N
PubChem CID 2777240
SMILES OC(=O)C1=C(O)C=CC(=C1)S(Cl)(=O)=O

Specifications

Melting Point 171°C to 174°C
Quantity 1 g
UN Number UN2923
Synonym 5-chlorosulfonyl-2-hydroxybenzoic acid, 5-chlorosulfonyl-2-hydroxy-benzoic acid, 5-chlorosulfonyl salicylic acid, benzoic acid, 5-chlorosulfonyl-2-hydroxy, benzoic acid,5-chlorosulfonyl-2-hydroxy, 5-chlorosulphonyl-2-hydroxybenzoic acid, 5-chlorosulfonyl-2-hydroxybenzoicacid, acmc-1cis5, 5-chlorosulphonyl salicylic acid
IUPAC Name 5-(chlorosulfonyl)-2-hydroxybenzoic acid
Formula Weight 236.63
Percent Purity 97%
Chemical Name or Material 5-(Chlorosulfonyl)salicylic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only