missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosalicylic Acid 98.0+%, TCI America™

Catalog No. p-7124337
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C028725G 25 g
C0287500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C028725G Supplier TCI America Supplier No. C028725G

Chemical Identifiers

CAS 321-14-2
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00002457
InChI Key NKBASRXWGAGQDP-UHFFFAOYSA-N
Synonym 5-chlorosalicylic acid, benzoic acid, 5-chloro-2-hydroxy, 2-hydroxy-5-chlorobenzoic acid, 5 csa, salicylic acid, 5-chloro, 5-chlorosalicyclic acid, 5-chloro salicylic acid, 5-chloro-2-hydroxy-benzoic acid, unii-j2yfg68vcz, ccris 4485
PubChem CID 9447
ChEBI CHEBI:420128
IUPAC Name 5-chloro-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(Cl)=CC=C1O

Specifications

Melting Point 173°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 172.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorosalicylic Acid
RTECSNumber VO2080000
TSCA Yes