missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chlorosalicylamide 98.0+%, TCI America™
Supplier: TCI America C078725G
Specifications
| 5-Chlorosalicylamide | |
| 234°C | |
| MFCD00007980 | |
| 5-chlorosalicylamide, chlorosalicylamide, benzamide, 5-chloro-2-hydroxy, 5-chlorosalicyl-amide, acmc-209ohl, 5chloro-2-hydroxy-benzamide, ksc498a5j, 5-chloro-2-hydroxy-benzamide, #, 5-chloranyl-2-oxidanyl-benzamide | |
| NC(=O)C1=CC(Cl)=CC=C1O | |
| 171.58 | |
| 171.58 | |
| Crystalline Powder |
| 7120-43-6 | |
| C7H6ClNO2 | |
| 25 g | |
| NNHMQZBVJPQCAK-UHFFFAOYSA-N | |
| 5-chloro-2-hydroxybenzamide | |
| 348094 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 7120-43-6 | |
| 171.58 | |
| NNHMQZBVJPQCAK-UHFFFAOYSA-N | |
| 348094 | |
| NC(=O)C1=CC(Cl)=CC=C1O |
| C7H6ClNO2 | |
| MFCD00007980 | |
| 5-chlorosalicylamide, chlorosalicylamide, benzamide, 5-chloro-2-hydroxy, 5-chlorosalicyl-amide, acmc-209ohl, 5chloro-2-hydroxy-benzamide, ksc498a5j, 5-chloro-2-hydroxy-benzamide, #, 5-chloranyl-2-oxidanyl-benzamide | |
| 5-chloro-2-hydroxybenzamide |
Safety and Handling
TSCA : No