missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-1,3-dimethyl-1h-pyrazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC12524DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC12524DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

852227-86-2
144.60
SGEZKPNUNBVVLB-UHFFFAOYSA-N
4961270
CN1N=C(C)C=C1CCl
C6H9ClN2
MFCD07368502
5-chloromethyl-1,3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dimethylpyrazole, 1h-pyrazole, 5-chloromethyl-1,3-dimethyl, 5-chloromethyl-1, pubchem23625, 3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dime..., 5-chloromethyl-1,4-dimethyl-1h-pyrazole, 1h-pyrazole,5-chloromethyl-1,3-dimethyl
5-(chloromethyl)-1,3-dimethylpyrazole

Specifications

5-(Chloromethyl)-1, 3-dimethyl-1H-pyrazole
97%
C6H9ClN2
1g
SGEZKPNUNBVVLB-UHFFFAOYSA-N
5-(chloromethyl)-1,3-dimethylpyrazole
4961270
97%
852227-86-2
Amber Glass Bottle
MFCD07368502
5-chloromethyl-1,3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dimethylpyrazole, 1h-pyrazole, 5-chloromethyl-1,3-dimethyl, 5-chloromethyl-1, pubchem23625, 3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dime..., 5-chloromethyl-1,4-dimethyl-1h-pyrazole, 1h-pyrazole,5-chloromethyl-1,3-dimethyl
CN1N=C(C)C=C1CCl
144.60
144.6
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

SDS
Documents

Documents