missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chlorobenzotriazole 98.0+%, TCI America™
$102.52 - $301.98
Chemical Identifiers
| CAS | 94-97-3 |
|---|---|
| Molecular Formula | C6H4ClN3 |
| Molecular Weight (g/mol) | 153.57 |
| MDL Number | MFCD00005700 |
| InChI Key | PZBQVZFITSVHAW-UHFFFAOYSA-N |
| Synonym | 5-chlorobenzotriazole, 1h-benzotriazole, 5-chloro, 6-chlorobenzotriazole, 5-chloro-1h-benzotriazole, 1h-benzotriazole, 6-chloro, 5-chloro-1h-benzo d 1,2,3 triazole, 5-chloro-1h-1,2,3-benzotriazole, 6-chloro-1h-benzotriazole, unii-8c7o46g78k, 5-chlorobenztriazole |
| PubChem CID | 66760 |
| IUPAC Name | 5-chloro-2H-1,2,3-benzotriazole |
| SMILES | ClC1=CC2=NNN=C2C=C1 |
Chemical Identifiers
| 94-97-3 | |
| 153.57 | |
| PZBQVZFITSVHAW-UHFFFAOYSA-N | |
| 66760 | |
| ClC1=CC2=NNN=C2C=C1 |
| C6H4ClN3 | |
| MFCD00005700 | |
| 5-chlorobenzotriazole, 1h-benzotriazole, 5-chloro, 6-chlorobenzotriazole, 5-chloro-1h-benzotriazole, 1h-benzotriazole, 6-chloro, 5-chloro-1h-benzo d 1,2,3 triazole, 5-chloro-1h-1,2,3-benzotriazole, 6-chloro-1h-benzotriazole, unii-8c7o46g78k, 5-chlorobenztriazole | |
| 5-chloro-2H-1,2,3-benzotriazole |
Specifications
| 94-97-3 | |
| White-Yellow | |
| MFCD00005700 | |
| 5-chlorobenzotriazole, 1h-benzotriazole, 5-chloro, 6-chlorobenzotriazole, 5-chloro-1h-benzotriazole, 1h-benzotriazole, 6-chloro, 5-chloro-1h-benzo d 1,2,3 triazole, 5-chloro-1h-1,2,3-benzotriazole, 6-chloro-1h-benzotriazole, unii-8c7o46g78k, 5-chlorobenztriazole | |
| ClC1=CC2=NNN=C2C=C1 | |
| 153.57 | |
| 153.57 | |
| Crystalline Powder |
| 158°C | |
| C6H4ClN3 | |
| 25 g | |
| PZBQVZFITSVHAW-UHFFFAOYSA-N | |
| 5-chloro-2H-1,2,3-benzotriazole | |
| 66760 | |
| ≥98.0% (GC,T) | |
| 5-Chlorobenzotriazole |
Safety and Handling
EINECSNumber : (5)-0540
TSCA : Yes