missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzofuran-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. C28901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28901G 1 g
C28905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28901G Supplier TCI America Supplier No. C28901G

Chemical Identifiers

CAS 10242-10-1
Molecular Formula C9H4ClO3
Molecular Weight (g/mol) 195.58
MDL Number MFCD00060512
InChI Key JETRXAHRPACNMA-UHFFFAOYSA-M
Synonym 5-chlorobenzofuran-2-carboxylic acid, 5-chlorobenzo b furan-2-carboxylic acid, 5-chloro-benzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 5-chloro, 2-benzofurancarboxylicacid, 5-chloro, 2-carboxy-5-chlorobenzo b furan, 5-chloro-2-benzofurancarboxylic acid, 5-chlorocoumarilic acid, acmc-20am44, 5-chlo-robenzofuran-2-carboxylic acid
PubChem CID 937838
IUPAC Name 5-chloro-1-benzofuran-2-carboxylate
SMILES [O-]C(=O)C1=CC2=CC(Cl)=CC=C2O1

Specifications

Melting Point 266°C
Color White-Yellow
Quantity 1 g
Formula Weight 196.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorobenzofuran-2-carboxylic Acid