accessibility menu, dialog, popup

UserName

OOOCl

CAS RN 10242-10-1

IUPAC Name: 5-chloro-1-benzofuran-2-carboxylate
Synonyms: 5-chloro-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 195.58
Molecular Formula: C9H4ClO3
InChi Key: JETRXAHRPACNMA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC(Cl)=CC=C2O1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)