missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzimidazole 98.0+%, TCI America™

Catalog No. C01305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01305G 5 g
C013025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C01305G Supplier TCI America Supplier No. C01305G
Only null left

Chemical Identifiers

CAS 4887-82-5
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.581
MDL Number MFCD00059698
InChI Key NKLOLMQJDLMZRE-UHFFFAOYSA-N
PubChem CID 78599
IUPAC Name 6-chloro-1H-benzimidazole
SMILES C1=CC2=C(C=C1Cl)NC=N2

Specifications

Melting Point 131°C
Quantity 5 g
Formula Weight 152.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorobenzimidazole