missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-o-cresol 97.0+%, TCI America™
$53.90 - $150.16
Chemical Identifiers
| CAS | 5306-98-9 |
|---|---|
| Molecular Formula | C7H7ClO |
| Molecular Weight (g/mol) | 142.58 |
| MDL Number | MFCD00060672 |
| InChI Key | KKFPXGXMSBBNJI-UHFFFAOYSA-N |
| PubChem CID | 79192 |
| IUPAC Name | 5-chloro-2-methylphenol |
| SMILES | CC1=CC=C(Cl)C=C1O |
Chemical Identifiers
| 5306-98-9 | |
| 142.58 | |
| KKFPXGXMSBBNJI-UHFFFAOYSA-N | |
| 5-chloro-2-methylphenol |
| C7H7ClO | |
| MFCD00060672 | |
| 79192 | |
| CC1=CC=C(Cl)C=C1O |
Specifications
| 5306-98-9 | |
| C7H7ClO | |
| 1 g | |
| 5-chloro-o-cresol, 5-chloro-2-cresol, o-cresol, 5-chloro, phenol, 5-chloro-2-methyl, 3-chloro-6-methylphenol, 2-methyl-5-chlorophenol, 4-chloro-2-hydroxytoluene, 5-chloro-2-methyl-phenol, phenol,5-chloro-2-methyl | |
| CC1=CC=C(Cl)C=C1O | |
| 142.58 | |
| 142.58 | |
| Crystalline Powder |
| 74°C | |
| MFCD00060672 | |
| 3437 | |
| KKFPXGXMSBBNJI-UHFFFAOYSA-N | |
| 5-chloro-2-methylphenol | |
| 79192 | |
| ≥97.0% (GC) | |
| 5-Chloro-o-cresol |
Safety and Handling
EINECSNumber : (3)-0900
TSCA : No